Pump tubing, PharMed® Ismaprene
Lieferant: Ismatec
PharMed®
ISMCSC0307AEA 52.4 EUR
ISMCSC0307A ISMCSC0305A ISMCSC0338A ISMCSC0303A ISMCSC0313A ISMC95809-32 ISMCSC0322A ISMCSC0341A ISMCSC0352A ISMCSC0348 ISMCSC0323 ISMCSC0344 ISMCSC0331 ISMCSC0353 224-2554 ISMCMF0013A
Pump tubing, PharMed® Ismaprene
Schläuche Schläuche für Peristaltikpumpen
Thermoplastic elastomer based on polypropylene, beige
  • Service life in peristaltic pumps is three times longer than that of comparable silicone tubing
  • Can be autoclaved numerous times
  • Conforms with the requirements of the US Pharmacopeia class VI, the US Food and Drug Administration (FDA) and the US National Sanitary Foundation (NSF)
Ideal for use in fermentation and separation, cell culture, syntheses and process monitoring. It is less permeable to gases and steam than silicon tubing. Good general chemical resistance, very good resistance to acids, alkalis and oxidants. Impermeable to white light and UV radiation, providing protection for sensitive liquids. Ideal for use with peristaltic pumps.
Temperature resistance: −60 to +135 °C
Complies with standards: FDA 21CFR177.2600/US Pharmacopoea Class VI/NSF listed (Standard 51).
Potential leaching of additives (lubricants).