Pump tubing, Tygon® MHSL 2001
Lieferant: Ismatec
Tygon®
ISMC06518-64EA 481 EUR
ISMC06518-64 ISMCSC0808A ISMC06518-22 ISMC06518-66 ISMCSC0804A ISMC06518-16 ISMC06518-08 ISMC06518-14 ISMC06518-04 ISMC96465-28 ISMC96466-14 ISMCSC0806 ISMCSC0818 ISMC06475-18 ISMC96466-42
Pump tubing, Tygon® MHSL 2001
Schläuche Schläuche für Peristaltikpumpen
The transparent, plasticiser-free tubing with superior pump-life. Eespecially designed for MEK and other aggressive solvents.
  • Plasticizer and oil-free
  • Smooth inner-bore
  • Low sorption maintains fluid and tube integrity
  • Does not impart anything into the pumping medium
  • No release of hazardous materials when properly incinerated
  • Complies with FDA standards
Temperature resistant from −73 to +57 °C
Hardness (Shore durometer A): 69
Density: 0,88