Pump tubing, Tygon® S3 E-LFL
Lieferant: Ismatec
ISMCSCE0409EA 92.6 EUR
ISMCSCE0409 ISMCSCE0408 ISMCSCE0418 ISMCSCE0407 ISMCSCE0390 224-2286 ISMCSCE0398 ISMCSCE0430 ISMCSCE0406 ISMCSCE0427 ISMCSCE0426 ISMCSCE0404 ISMCSCE0403 ISMCSCE0425 ISMCSCE0414 ISMCSCE0424 ISMCSCE0401 ISMCSCE0423 ISMCSCE0411 ISMCSCE0400 ISMCSCE0422 ISMCSCE0421 ISMCSCE0399 ISMCSCE0410
Pump tubing, Tygon® S3 E-LFL
Schläuche Schläuche für Peristaltikpumpen
Non DEHP and phthalate-free tubing.
  • Over 1000 hours pump tubing life at 0 psi.; over 600 hours pump tubing life at 10 psi.
  • Extremely low particle spallation
  • Contains no BPA or phthalates
  • Broad chemical resistance
  • Meets USP Class VI, ISO 10993 and FDA criteria - suitable for pharma, medical and food applications
Longest flexible life of any clear Tygon® tubing. The non ageing and broad chemical resistance properties enable users to utilise the tubing for transferring chemicals, cleaning agents and bactericides without danger of corrosion (as with metals) or oxidation (as with rubber). Tygon® S3™ E-LFL tubing is formulated specifically for use in peristaltic pump applications.
Temperature resistant from −55 to +74 °C
Hardness (Shore durometer A): 56
Density: 1,17