Kieselgel, Porengröße: 60 Å
BKEMSC-9982-B010EA
63
EUR
BKEMSC-9982-B010
Kieselgel, Porengröße: 60 Å
Silica gel
Formel:
SiO₂ Molecular Weight: 60,08 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011232 CAS-Nummer: 63231-67-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...