5-Aminoisatosäureanhydrid
Lieferant: FLUOROCHEM LTD (CHEMPUR)
FLUO018184-5GEA
383
EUR
FLUO018184-5G
FLUO018184-1G
5-Aminoisatosäureanhydrid
5-Aminoisatosäureanhydrid
Formel:
C₈H₆N₂O₃ Molecular Weight: 178,15 g/mol |
MDL:
MFCD00060534 CAS-Nummer: 169037-24-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...