Cyclohexancarbonitril

Lieferant: CHEMPUR

CHMPRH01750.250EA 0 EUR
CHMPRH01750.250 MAYBRH01750.1
Cyclohexancarbonitril
Cyclohexancarbonitril
Formel: C₇H₁₁N
Molecular Weight: 109,17 g/mol
MDL: MFCD00040444
CAS-Nummer: 766-05-2
EINECS: 212-157-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR