Phenyltrifluorsilan

Lieferant: SILANE (PART OF CHEMPUR)

SILNSILS13725.10EA 0 EUR
SILNSILS13725.10 SILNSILS13725.25 SILNSILS13725.100
Phenyltrifluorsilan
Phenyltrifluorosilane
Formel: C₆H₅F₃Si
CAS-Nummer: 368-47-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR