Chlor(diisopropyl)phosphin

Lieferant: CHEMPUR

CHMPFL13476.POREA 0 EUR
CHMPFL13476.POR
Chlor(diisopropyl)phosphin
Chlor(diisopropyl)phosphin
Formel: C₆H₁₄ClP
Molecular Weight: 152,6 g/mol
MDL: MFCD00015027
CAS-Nummer: 40244-90-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR