Diisodecylphthalat

Lieferant: EHRENSTORFER GMBH

EHERC16173550EA 51 EUR
EHERC16173550
Diisodecylphthalat
Diisodecylphthalat
Formel: C₂₈H₄₆O₄
Molecular Weight: 446,67 g/mol
MDL: MFCD00048389
CAS-Nummer: 26761-40-0
EINECS: 247-977-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR