Diallyldiphenylsilan

Lieferant: SILANE (PART OF CHEMPUR)

SILNSILS07775.5EA 0 EUR
SILNSILS07775.5 SILNSILS07775.25
Diallyldiphenylsilan
Diallyldiphenylsilan
Formel: C₁₈H₂₀Si
Molecular Weight: 264,44 g/mol
MDL: MFCD00026068
CAS-Nummer: 10519-88-7
EINECS: 234-061-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR