1,4,7,10-Tetraazacyclododecan (Cyclen, Kryptofix® 11 aza) ≥95%

Lieferant: ABCR GMBH

ABCRAB127386.0025EA 0 EUR
ABCRAB127386.0025
1,4,7,10-Tetraazacyclododecan (Cyclen, Kryptofix® 11 aza) ≥95%
1,4,7,10-Tetraazacyclododecan (Cyclen, Kryptofix® 11 aza)
Formel: C₈H₂₀N₄
Molecular Weight: 172,27 g/mol
MDL: MFCD00066281
CAS-Nummer: 294-90-6
EINECS: 425-450-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR