4-Nitro-2-(trifluormethyl)benzonitril, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL579955-5GEA
67.6
EUR
SIAL579955-5G
4-Nitro-2-(trifluormethyl)benzonitril, Sigma-Aldrich®
4-Nitro-2-(trifluormethyl)benzonitril
Formel:
C₈H₃F₃N₂O₂ Molecular Weight: 216,12 g/mol |
MDL:
MFCD01075729 CAS-Nummer: 320-47-8 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...