2,3,7-Trimethylindol

Lieferant: CHEMPUR

CHMPFL13736.POREA 0 EUR
CHMPFL13736.POR
2,3,7-Trimethylindol
2,3,7-Trimethylindole
Formel: C₁₁H₁₃N
CAS-Nummer: 27505-78-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR