1,2,3,4,5-Pentabrom-6-chlorbenzol
Lieferant: Thermo Scientific
MAYBBTB15046.250EA
0
EUR
MAYBBTB15046.250
MAYBBTB15046.1
1,2,3,4,5-Pentabrom-6-chlorbenzol
1,2,3,4,5-Pentabrom-6-chlorbenzol
Formel:
C₆Br₅Cl |
CAS-Nummer:
13075-05-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...