1-(4-Bromphenyl)piperazin, Sigma-Aldrich®

Lieferant: Merck

SIAL724580-1GEA 33.9 EUR
SIAL724580-1G
1-(4-Bromphenyl)piperazin, Sigma-Aldrich®
1-(4-Bromphenyl)piperazin
Formel: C₁₀H₁₃BrN₂
Molecular Weight: 241,13 g/mol
MDL: MFCD00130198
CAS-Nummer: 66698-28-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR