1,2-Epoxytetradecan, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Dodecyloxiran
M-Clarity™ quality level = MQ100
SIAL260266-100MLEA
158
EUR
SIAL260266-100ML
1,2-Epoxytetradecan, Sigma-Aldrich®
1,2-Epoxytetradecan
Formel:
C₁₄H₂₈O Molecular Weight: 212,38 g/mol Siedepunkt: 95 °C (1013 hPa) Dichte: 0,845 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00037524 CAS-Nummer: 3234-28-4 EINECS: 221-781-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...