2,3,4-Trichloranisol

Lieferant: EHRENSTORFER GMBH

EHERC17713400EA 39 EUR
EHERC17713400
2,3,4-Trichloranisol
2,3,4-Trichloranisol
Formel: C₇H₅Cl₃O
Molecular Weight: 211,47 g/mol
CAS-Nummer: 54135-80-7
EINECS: 258-990-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR