Safety syringes, BD Integra™
Lieferant: BD Medical
BD Integra™
BDAM305269EA 131 EUR
BDAM305269
Safety syringes, BD Integra™
This safety syringe features a retracting mechanism which allows for the device to activate immediately and completely shields the used needle, quickly eliminating the risk of exposure to a needlestick injury.
Detachable needle provides the benefit of low waste space, which is important in reducing medication waste and costs.