4-Nitrodiphenylamin

Lieferant: EHRENSTORFER GMBH

Synonyme: 4-Nitro-N-phenylanilin

EHERC15559400EA 65 EUR
EHERC15559400
4-Nitrodiphenylamin
4-Nitrodiphenylamin
Formel: C₁₂H₁₀N₂O₂
Molecular Weight: 214,22 g/mol
MDL: MFCD00007301
CAS-Nummer: 836-30-6
EINECS: 212-646-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR