Isobutyraldehyd ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-Methylpropanal
A12106.0FEA
72.8
EUR
A12106.0F
A12106.AP
A12106.PG
Isobutyraldehyd ≥98%
Isobutyraldehyd
Purity: 98%
CAS No.: 78-84-2
Molecular Formula: C4H8O
Formula Weight: 72.11
Storage Temperature: 2°C to 8°C
Flash Point: -24°(-11°F)
Physical Form: Liquid
Density: 0.794
Refractive Index: 1.3723
Sensitivity: Air Sensitive
Appearance: Colourless
Odour: Pungent
Melting Point: -65°
Boiling Point: 63-65°
Beilstein Reference: 605330
Merck Reference: 14,5154
UN No.: 2045
MDL No.: MFCD00006980
Formel:
C₄H₈O Molecular Weight: 72,11 g/mol Siedepunkt: 63…65 °C Schmelzpunkt: –65 °C Dichte: 0,794 Flammpunkt: –24 °C (–11 °F) Lagertemperatur: Kühlschrank |
MDL:
MFCD00006980 CAS-Nummer: 78-84-2 EINECS: 201-149-6 UN: 2045 ADR: 3,II Merck Index: 13,05171 |