n-Octatriacontan
Lieferant: EHRENSTORFER GMBH
EHERC15711280EA
74
EUR
EHERC15711280
n-Octatriacontan
n-Octatriacontan
Formel:
H₃C(CH₂)₃₆CH₃ Molecular Weight: 535,04 g/mol Schmelzpunkt: 79 °C Lagertemperatur: Raumtemperatur |
CAS-Nummer:
7194-85-6 EINECS: 230-560-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...