3-Brom-5-fluorbenzaldehyd

Lieferant: CHEMPUR

CHMPFL13207.POREA 0 EUR
CHMPFL13207.POR
3-Brom-5-fluorbenzaldehyd
3-Bromo-5-fluorobenzaldehyde
Formel: C₇H₄BrFO
MDL: MFCD04116319
CAS-Nummer: 188813-02-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR