Baicalein

Lieferant: Molekula

Synonyme: 5,6,7-Trihydroxy-2-phenylchromen-4-on5,6,7-Trihydroxyflavon

MOLEM21479410EA 0 EUR
MOLEM21479410
Baicalein
Baicalein
Formel: C₁₅H₁₀O₅
Molecular Weight: 270,24 g/mol
MDL: MFCD00017459
CAS-Nummer: 491-67-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR