2-(Trifluormethyl)benzolsulfonamid ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-(Trifluormethyl)benzolsulfonsäureamid
, α,α,α-Trifluor-o-toluolsulfonamid
L19717.03EA
67.9
EUR
L19717.03
2-(Trifluormethyl)benzolsulfonamid ≥97%
2-(Trifluormethyl)benzolsulfonamid
Formel:
C₇H₆F₃NO₂S Molecular Weight: 225,19 g/mol Schmelzpunkt: 188…192 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042420 CAS-Nummer: 1869-24-5 UN: 2811 ADR: 6.1,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...