Prednison

Lieferant: Molekula

MOLE27214047-5GEA 140 EUR
MOLE27214047-5G
Prednison
Prednison
Formel: C₂₁H₂₆O₅
Molecular Weight: 358,43 g/mol
Schmelzpunkt: 230…232 °C
MDL: MFCD00003608
CAS-Nummer: 53-03-2
EINECS: 200-160-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR