VWR®, Reagenzglas-, Röhrchen- und Abtropfgestelle
Lieferant: VWR
VWR®
211-0222EA 12.7 EUR
211-0222
VWR®, Reagenzglas-, Röhrchen- und Abtropfgestelle
Gestelle Reagenzglasgestelle
PP, verstärkt.
  • Heat resistant up to 135 °C, autoclavable
  • Do not float in a water bath
  • Excellent chemical resistance
Secure hold for test tubes, sample tubes and reaction vessels. Can be used as drying racks for test tubes, electrophoresis and chromatography plates.