Cell culture plates, poly-D-lysine coated
Lieferant: Corning Life Sciences
Corning®
734-2292EA 1871.47 EUR
734-2292 734-2294 734-2293 734-2291 CORN3768
Cell culture plates, poly-D-lysine coated
Mikrotiterplatten Zellkultur-Platten Zellkulturplatten, beschichtet
PS, flat bottom, sterile (aseptic assembly) and certified non-pyrogenic, with lid. Microplates coated with poly-D-lysine for enhanced cell attachment.
  • Non reversible lids with condensation rings to reduce evaporation (96-well plates)
  • Optically clear flat well bottom permits direct microscopic viewing
  • Opaque walls to prevent well-to-well crosstalk
  • Can be used for both top and bottom reading instruments
  • Individual alphanumeric codes for well identification
Microplates coated with poly-D-lysine for enhanced cell attachment.
  • Non reversible lids with condensation rings to reduce evaporation (96-well plates)
  • Optically clear flat well bottom permits direct microscopic viewing
  • Opaque walls to prevent well-to-well crosstalk
  • Can be used for both top and bottom reading instruments
  • Individual alphanumeric codes for well identification
Verpackung: Bulk packed, 20 per bag.