Strip plates, 96-well, Corning® Stripwell™
Lieferant: Corning
Corning® Stripwell®
444-1021EA 840.27 EUR
444-1021 732-5501
Strip plates, 96-well, Corning® Stripwell™
Mikrotiterplatten Immunologie-Mikrotiterplatten
PS, flat bottom. Designed for in vitro diagnostic assays. The flat bottom strips easily break apart and are pre-assembled in an 'egg-crate' style strip holder that allows each individual well to be positioned back into the plate once broken.
  • Available as low volume or standard format with well volumes of 190 µl (working volume 75 to 125 µl) and 360 µl (working volume 75 to 200 µl) respectively
  • 1×8 strips are designed to fit only one way into the strip holder, eliminating the chance of misorientation
*Product has a certified surface chemistry
**Individual 1×8 strips without frame, bulk packed
***Microplates individually packaged with lid