Natriumstannat SnO₂ 42-45%
Lieferant: Honeywell Chemicals
|
Achtung
|
Synonyme:
Dinatriumzinntrioxid
, Natriumzinn(IV)oxid
HONE14505.9050EA
0
EUR
HONE14505.9050
Natriumstannat SnO₂ 42-45%
Natriumstannat
Formel:
Na₂O₃Sn Molecular Weight: 212,69 g/mol Dichte: 4,68 g/cm³ (25 °C) |
MDL:
MFCD00014241 CAS-Nummer: 12058-66-1 EINECS: 235-030-5 Merck Index: 12,08826 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...