3,5-Di-tert-butylbrenzcatechin, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ quality level = MQ100
SIALD45800-25GEA
80.9
EUR
SIALD45800-25G
3,5-Di-tert-butylbrenzcatechin, Sigma-Aldrich®
3,5-Di-tert-butylbrenzcatechin
Formel:
C₁₄H₂₂O₂ Molecular Weight: 222,33 g/mol Siedepunkt: 310 °C (1013 hPa) Schmelzpunkt: 96…97 °C |
MDL:
MFCD00008819 CAS-Nummer: 1020-31-1 EINECS: 213-816-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...