Dichlor(methyl)silan
Lieferant: SILANE (PART OF CHEMPUR)
Synonyme:
Methyldichlorsilan
SILNSILS11200.25EA
0
EUR
SILNSILS11200.25
SILNSILS11200.100
Dichlor(methyl)silan
Dichlor(methyl)silan
Formel:
CH₄Cl₂Si Molecular Weight: 115,03 g/mol Siedepunkt: 41 °C (1013 hPa) Schmelzpunkt: –92,8 °C Dichte: 0,93 g/cm³ (0 °C) |
MDL:
MFCD00000494 CAS-Nummer: 75-54-7 EINECS: 200-877-1 UN: 1242 ADR: 4.3,I REACH: 01-2119436669-21 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...