Benzyltrichlorsilan
Lieferant: SILANE (PART OF CHEMPUR)
SILNSILS01000.10EA
0
EUR
SILNSILS01000.10
SILNSILS01000.100
SILNSILS01000.25
Benzyltrichlorsilan
Benzyltrichlorsilan
Formel:
C₇H₇Cl₃Si |
CAS-Nummer:
770-10-5 EINECS: 212-219-0 UN: 2987 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...