Allyltrimethylsilan
Lieferant: SILANE (PART OF CHEMPUR)
SILNSILS00450.25EA
76.5
EUR
SILNSILS00450.25
SILNSILS00450.100
SILNSILS00450.10
Allyltrimethylsilan
Allyltrimethylsilan
Formel:
H₂CCHCH₂Si(CH₃)₃ Molecular Weight: 114,26 g/mol Siedepunkt: 84 °C (1013 hPa) Dichte: 0,7139 g/cm³ (20 °C) |
MDL:
MFCD00008635 CAS-Nummer: 762-72-1 EINECS: 212-104-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...