4-Amino-3-fluorpyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL704490-1GEA 86.2 EUR
SIAL704490-1G
4-Amino-3-fluorpyridin, Sigma-Aldrich®
4-Amino-3-fluoropyridine
Formel: C₅H₅FN₂
Molecular Weight: 112,11 g/mol
MDL: MFCD00234072
CAS-Nummer: 2247-88-3
UN: 2811
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR