4-Amino-2-chlorpyrimidin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Chlorpyrimidin-4-ylamin
M-Clarity™ quality level = MQ100
SIAL679062-1GEA
57
EUR
SIAL679062-1G
4-Amino-2-chlorpyrimidin, Sigma-Aldrich®
4-Amino-2-chloropyrimidine
Formel:
C₄H₄ClN₃ Molecular Weight: 129,55 g/mol |
MDL:
MFCD00194051 CAS-Nummer: 7461-50-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...