4-(Trifluormethyl)pyridin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ200
SIAL522910-1GEA
28.1
EUR
SIAL522910-1G
4-(Trifluormethyl)pyridin, Sigma-Aldrich®
4-(Trifluoromethyl)pyridine
Formel:
C₆H₄F₃N Molecular Weight: 147,1 g/mol Siedepunkt: 105…106 °C (1013 hPa) |
MDL:
MFCD00153289 CAS-Nummer: 3796-24-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...