5-Indolcarbaldehyd, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Indol-5-carboxaldehyd
M-Clarity™ quality level = MQ100
SIAL513830-5GEA
41.6
EUR
SIAL513830-5G
5-Indolcarbaldehyd, Sigma-Aldrich®
5-Indolcarbaldehyd
Formel:
C₉H₇NO Molecular Weight: 145,16 g/mol |
MDL:
MFCD02093664 CAS-Nummer: 1196-69-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...