tert-Butylcyanacetat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Cyanessigsäure-tert-butylester
SIAL28440-10ML-FEA
58.5
EUR
SIAL28440-10ML-F
tert-Butylcyanacetat, Sigma-Aldrich®
tert-Butylcyanacetat
Formel:
C₇H₁₁NO₂ Molecular Weight: 141,17 g/mol Dichte: 0,987 g/cm³ (20 °C) |
MDL:
MFCD00001938 CAS-Nummer: 1116-98-9 EINECS: 214-243-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...