5-Fluor-2-indolcarbonsäure, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ100
SIAL265128-5GEA
303
EUR
SIAL265128-5G
5-Fluor-2-indolcarbonsäure, Sigma-Aldrich®
5-Fluoroindole-2-carboxylic acid
Formel:
C₉H₆FNO₂ Molecular Weight: 179,15 g/mol Schmelzpunkt: 258…259 °C |
MDL:
MFCD00005612 CAS-Nummer: 399-76-8 EINECS: 206-919-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...