Wasser Reag. Ph. Eur. 1095507 partikelfrei
Lieferant: VWR Chemicals
Synonyme: WasserstoffoxidHydrogenoxid
87962.290EA 59.4 EUR
87962.290
Wasser Reag. Ph. Eur. 1095507 partikelfrei
Wasser
Bestellinformation: Geliefert mit Analysenzertifikat.
Formel: H₂O
Molecular Weight: 18,02 g/mol
Siedepunkt: 100 °C (1013 hPa)
Schmelzpunkt: 0 °C
Dichte: 1 g/cm³ (4 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 12,10175
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022
Bestellen
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022