9-Fluorencarbonsäure
Lieferant: Molekula
MOLE17538892-25GEA
0
EUR
MOLE17538892-25G
9-Fluorencarbonsäure
9-Fluorencarbonsäure
Formel:
C₁₄H₁₀O₂ Molecular Weight: 210,23 g/mol Siedepunkt: 365 °C (1013 hPa) Schmelzpunkt: 226…229 °C Dichte: 1,333 g/cm³ (20 °C) |
MDL:
MFCD00001136 CAS-Nummer: 1989-33-9 EINECS: 217-866-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...