4,5-Dimethylthiazol

Lieferant: Molekula

MOLE11765967-25GEA 0 EUR
MOLE11765967-25G
4,5-Dimethylthiazol
4,5-Dimethylthiazole
Formel: C₅H₇NS
Molecular Weight: 113,18 g/mol
Siedepunkt: 149…153 °C (1013 hPa)
Dichte: 1,0828 g/cm³ (20 °C)
MDL: MFCD00005336
CAS-Nummer: 3581-91-7
EINECS: 222-703-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR