1,3-Dimethyluracil

Lieferant: Molekula

Synonyme: 2,4-Dihydroxy-1,3-dimethylpyrimidin

MOLE25242304-5GEA 0 EUR
MOLE25242304-5G
1,3-Dimethyluracil
1,3-Dimethyluracil
Formel: C₆H₈N₂O₂
Molecular Weight: 140,14 g/mol
MDL: MFCD00038065
CAS-Nummer: 874-14-6
EINECS: 212-856-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR