4-Amino-2,6-dichlorpyrimidin

Lieferant: Molekula

MOLE20263504-1GEA 0 EUR
MOLE20263504-1G
4-Amino-2,6-dichlorpyrimidin
4-Amino-2,6-dichloropyrimidine
Formel: C₄H₃Cl₂N₃
Molecular Weight: 163,99 g/mol
MDL: MFCD00038015
CAS-Nummer: 10132-07-7
EINECS: 233-369-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR