1-(2,6-Diethylphenyl)-2-thioharnstoff
Lieferant: MENAI (PART OF CHEMPUR)
MENADH657.25EA
0
EUR
MENADH657.25
MENADH657.50
1-(2,6-Diethylphenyl)-2-thioharnstoff
1-(2,6-Diethylphenyl)-2-thioharnstoff
Formel:
(C₆H₃(CH₂CH₃)₂)NC(S)NH₂ Molecular Weight: 208,33 g/mol Siedepunkt: 336 °C (1013 hPa) Schmelzpunkt: 196 °C |
MDL:
MFCD00041166 CAS-Nummer: 25343-30-0 UN: 2811 ADR: 6.1,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...