Volasil 0244 volatile, klar, farblos
|
Achtung
|
633004VEA
0
EUR
633004V
Volasil 0244 volatile, klar, farblos
Octamethylcyclotetrasiloxan
Low toxicity, low odour solvent for a wide range of organic materials
- Ozonfreundlich
Formel:
[-Si(CH₃)₂O-]₄ Molecular Weight: 296,62 g/mol Siedepunkt: 175 °C (1013 hPa) Schmelzpunkt: 18 °C Dichte: 0,95 g/cm³ (20 °C) Flammpunkt: 51 °C |
MDL:
MFCD00003269 CAS-Nummer: 556-67-2 EINECS: 209-136-7 UN: 1993 ADR: 3,III Merck Index: 12,06843 |
Spezifikation Testergebnisse
Appearance Spec | Clear fluid free from extraneous matter |
Water | <75 ppm |
Content - D4 | 96.00 - 100.00% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...