1-(2-Chlor-6-fluorbenzyl)piperazin
Lieferant: Thermo Scientific
MAYBAW00532.250EA
0
EUR
MAYBAW00532.250
MAYBAW00532CB-250M
1-(2-Chlor-6-fluorbenzyl)piperazin
1-(2-Chlor-6-fluorbenzyl)piperazin
Formel:
C₁₁H₁₄ClFN₂ Molecular Weight: 228,7 g/mol |
MDL:
MFCD00816605 CAS-Nummer: 215655-20-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...