4,5-Dichlor-o-phenylendiamin
Lieferant: Thermo Scientific
Synonyme:
1,2-Diamino-4,5-dichlorbenzol
, 4,5-Dichlorbenzol-1,2-diamin
, 4,5-Dichlor-1,2-phenylendiamin
MAYB02-7389EA
0
EUR
MAYB02-7389
4,5-Dichlor-o-phenylendiamin
4,5-Dichlor-o-phenylendiamin
Formel:
C₆H₆Cl₂N₂ Molecular Weight: 177,03 g/mol Siedepunkt: 305 °C (1013 hPa) Schmelzpunkt: 163 °C |
MDL:
MFCD00007723 CAS-Nummer: 5348-42-5 EINECS: 226-305-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...