5-Fluor-2-nitrobenzoesäure ≥97%

Lieferant: Thermo Scientific
Achtung

B23133.06EA 83.9 EUR
B23133.06
5-Fluor-2-nitrobenzoesäure ≥97%
5-Fluor-2-nitrobenzoesäure
Formel: C₇H₄FNO₄
Molecular Weight: 185,11 g/mol
Schmelzpunkt: 134…135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00055635
CAS-Nummer: 320-98-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR