4-Amino-2,6-dichlorpyrimidin ≥98%

Achtung

A17294.06EA 57.3 EUR
A17294.06
4-Amino-2,6-dichlorpyrimidin ≥98%
4-Amino-2,6-dichloropyrimidine
Formel: C₄H₃Cl₂N₃
Molecular Weight: 163,99 g/mol
Schmelzpunkt: 272…274 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00038015
CAS-Nummer: 10132-07-7
EINECS: 233-369-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR